Learn More
Brady™ Oil Only Pillows
Catalog No. 19000169 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
19 gal./Bale
28 gal./Bale
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-000-169 19 gal./Bale
19-000-168 28 gal./Bale
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-000-169 Supplier Brady™ Supplier No. OIL99
For absorbing oil and petroleum based spills and leaking fluids in tight spaces
  • Feature white spunbond outer covering filled with white polypropylene
  • For use in waste troughs, coolant tanks, fluid reservoirs and sump wells
  • Specifications
    Capacity (English) 19 gal./Bale
    Type Oil Only Pillow
    For Use With (Application) Overspray leaks and drips, drums - liquid storage, drains
    Packaging Quantity 32/Cs.