Learn More
Brady™ Re-Form™ Multiwik™ Socks
Catalog No. 19000181 Shop All Brady Corporation Products
Change view
Click to view available options
Packaging Quantity:
4/Cs.
40/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
19-000-181 40/Cs.
19-000-180 4/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-000-181 Supplier Brady™ Supplier No. MS50
  • $240.00
    $167.65 / Case of 40
    Save $72.35
    30% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Dependable, flexible tubes that contain and absorb liquids
  • Molds around corners, and conform to uneven surface
  • Wringable and incinerable
  • Not for use with aggressive acids and caustics
  • Specifications
    Material Recycled Newsprint, Raw Cellulose
    Type Re-Form Multiwik Socks
    Packaging Quantity 40/Cs.