Learn More
Aviva Systems Biology Corp 4933434E20Rik Antibody - N-terminal region (ARP54536_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54536_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against 4933434E20Rik. It was validated on Western Blot. . Alternate names: NICE-3, NS5ATP4, AI462154, 5730552F22RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-936