Learn More
Scott Safety™ ACSi SCBA
Catalog No. 19122029
Change view
Click to view available options
Facepiece Type:
AV-3000 SureSeal (M) Poly
Catalog No. Facepiece Type
19-122-029 AV-3000 SureSeal (M) Poly
1 options
Catalog No. 19-122-029 Supplier Scott Safety™ Supplier No. 7012623344
Please contact your local Fisher Scientific sales representative for pricing
Offers the Advanced Carrying System (ACS), which features a padded Thermoflex backplate molded to fit the spine and back muscles for increased comfort and flexibility.
  • Lightweight, flexible Thermoflex backplate for greater comfort and flexibility
  • EZ-Flo regulator reduces inhalation and exhalation resistance for reduced user fatigue
  • Pneumatics are easily removed with a Scott “quarter-turn”—no tools required—for ease of maintenance and turn-around
  • Adjustable, padded shoulder straps and waistband with quick-release buckle for comfort and ease of donning and doffing
  • Shoulder mounted photoluminescent gauge with whistle end-of-service alarm for ease of use
  • Cylinder: 2216 psi Aluminum, 30 min.
Specifications
Product Type Industrial SCBA
Pressure 2216 psi
Facepiece Type AV-3000 SureSeal (M) Poly
Harness Style ACSI Standard
Hose Type None
Includes Cylinder: 2216 psi Aluminum, 30 min.; Manifold: Standard
Model ACS3A2013006101
Regulator Type EZ-Flo Standard
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.