Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Non-Combustible Outdoor Safety Lockers

Catalog No. 19414016
Change view
Click to view available options
Dimensions (D x W x H) Interior:
30 x 54 x 55.5 in.
54 x 54 x 62 in.
54 x 78 x 84 in.
78 x 78 x 84 in.
102 x 78 x 84 in.
102 x 102 x 84 in.
Explosion Relief Panel:
No
Yes
Load Bearing Capacity:
12 x 55 gal. drums
16 x 55 gal. drums
2 x 55 gal. drums
4 x 55 gal. drums
6 x 55 gal. drums
9 x 55 gal. drums
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H) Interior Explosion Relief Panel Load Bearing Capacity
19-414-016 102 x 102 x 84 in. Yes 16 x 55 gal. drums
19-414-005 30 x 54 x 55.5 in. No 2 x 55 gal. drums
19-414-006 30 x 54 x 55.5 in. Yes 2 x 55 gal. drums
19-414-007 54 x 54 x 62 in. No 4 x 55 gal. drums
19-414-008 54 x 54 x 62 in. Yes 4 x 55 gal. drums
19-414-009 54 x 78 x 84 in. No 6 x 55 gal. drums
19-414-010 54 x 78 x 84 in. Yes 6 x 55 gal. drums
19-414-011 78 x 78 x 84 in. No 9 x 55 gal. drums
19-414-012 78 x 78 x 84 in. Yes 9 x 55 gal. drums
19-414-013 102 x 78 x 84 in. No 12 x 55 gal. drums
19-414-014 102 x 78 x 84 in. Yes 12 x 55 gal. drums
19-414-015 102 x 102 x 84 in. No 16 x 55 gal. drums
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 19-414-016 Supplier Justrite™ Supplier No. 911161
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for storage of flammable and combustible liquids contained in chemical drums varying in capacities

  • Made of heavy gauge steel (backed by 15 year structural warranty)
  • Leakproof all welded sump secondary containment
  • Removable, galvanized steel grating
  • Three-point keyed locking hardware
  • Natural draft ventilation
  • Multiple static grounding connections
  • Anchoring plates for maximum hold down protection
  • Chemical and UV resistant coating, inside and out
  • Forklift channels for easy relocation
  • Available with and without Explosion Relief Panels
  • NFPA 704 diamond decal provides Right to Know information
  • FM-approved - including sump capacity and wind/snow load requirements

Model Specific Features:

  • With Explosion relief panels designed to release at 20 psf (911021, 911041, 911061, 911091, 911121, and 911161)
  • Without Explosion relief panels (911020, 911040, 911060, 911090, 911120 and 911160)

Certifications

FM approved

Compliance

Meets EPA, NFPA 30 and NFPA 1 requirements

Specifications

Product Type Non-combustible Outdoor Safety Locker
Explosion Relief Panel Yes
Dimensions (D x W x H) Interior 102 x 102 x 84 in.
Depth (Metric) Exterior 279.4 cm
Height (Metric) Interior 213.4 cm
No. of Doors 1
Depth (Metric) Interior 259.1 cm
Width (Metric) Exterior 292.1 cm
Load Bearing Capacity 16 x 55 gal. drums
Width (Metric) Interior 259.1 cm
Height (Metric) Exterior 246.4 cm
For Use With (Application) Hazardous/chemical storage
Color Bone
Description Heavy gauge steel
Width (English) Exterior 115 in.
Material Steel
Door Style Three-point lock
Width (English) Interior 102 in.
Height (English) Exterior 97 in.
Depth (English) Interior 102 in.
Depth (English) Exterior 110 in.
Certifications/Compliance FM approved, EPA, NFPA 30, NFPA 1 compliant
Height (English) Interior 84 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.