Learn More
Scott Safety™ AIR-PAK 75i SCBAs
Catalog No. 19122018
Change view
Click to view available options
Facepiece Type:
AV-3000 Sureseal Dupont™ Kevlar™ aramid fibers (M)
No Facepiece
Regulator Type:
E-Flo regulator w/Vibralert Quick Connect
No regulator
Pressure:
4500 psi
Includes:
Airline option, Hansen HK socket
No airline option
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Facepiece Type Regulator Type Pressure Includes
19-122-018 No Facepiece No regulator 4500 psi No airline option
19-122-015 AV-3000 Sureseal Dupont™ Kevlar™ aramid fibers (M) E-Flo regulator w/Vibralert Quick Connect 4500 psi Airline option, Hansen HK socket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-122-018 Supplier Scott Safety™ Supplier No. 7012623370
Please contact your local Fisher Scientific sales representative for pricing
Designed with durability and dependability for industrial applications.
  • Streamlined backframe provides comfortable fit with lower profile
  • Featues one-piece backframe constructed of lightweight, time-tested, durable aluminum alloy
  • Parachute-Style, oversized harness fasteners designed with gloved hands
  • Enhanced latching system with durable para-aramid fabric band
  • Includes efficient breathing circuit
  • Features Top Down Convertibility concept for c increase in versatility, compatibility with Scott product line
  • E-Z Flo regulator with Vibralert end-of-service indicator
  • Has two dual-redundant pressure reducer interconnected through automatic transfer valve
  • Quick-don mounting system in E-Z Flo regulator
  • Air-saver switch angled for easy operation with gloved hand
  • Fully padded shoulder straps and waist belt
  • Integrated donning/carrying handles
  • Dual mechanical speaking diaphragms
  • Remote pressure gauge
  • Cam-Over center cylinder adjustment band
  • Drag rescue loop
Specifications
Product Type SCBA
Pressure 4500 psi
Facepiece Type No Facepiece
For Use With (Application) Industrial applications
Includes No airline option
Model API454001000101
Regulator Type No regulator
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.