Learn More
MSC Loctite 135467 1 oz Bottle Clear Instant Adhesive Series 495, 20 sec Fixture Time, 24 hr Full Cure Time, Bonds to Metal, Plastic & Rubber
Supplier:  MSC 88545629
Encompass
Loctite 135467 1 oz Bottle Clear Instant Adhesive Series 495, 20 sec Fixture Time, 24 hr Full Cure Time, Bonds to Metal, Plastic & Rubber
Catalog No. 50-111-3865