Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Native pepsin from Porcine Stomach Mucosa

Catalog No. 516360500MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-636-0500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 51-636-0500MG Supplier MilliporeSigma™ Supplier No. 516360500MG
Add to Cart
Add to Cart

Catalyzes hydrolysis of aminoacyl-proline to amino acid and proline

  • Secreted by mucosal lining of stomach
  • Active under acidic conditions
  • Inhibitors include aliphatic alcohols, pepstatin A, and pH: >6.0
  • Lyophilized
  • Molar mass: 35,000g/mole
  • pH: 2 - 4

TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 35,000g/mole
Name Native pepsin
pH Range 2 - 4
Quantity 500 mg
Source Porcine stomach mucosa
Storage Requirements -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.