Learn More
Abbott™ iScreen™ External Drug Controls
Catalog No. 22911083
Change view
Click to view available options
Quantity:
5 mL
Type:
Drug Control Cotinine Specific 2X Positive
Catalog No. Quantity Type
22-911-083 5 mL Drug Control Cotinine Specific 2X Positive
1 options
This item is not returnable. View return policy
Catalog No. 22-911-083 Supplier Abbott™ Supplier No. 19000102
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Used as quality control urine to monitor precision of laboratory urine toxicology testing procedures for different analytes
Control designed to work with any urine drug screens
Specifications
Type Drug Control Cotinine Specific 2X Positive
Intended Use Used as quality control urine to monitor precision of laboratory urine toxicology testing procedures
Quantity 5 mL