Learn More
Abbott™ Integrated E-Z Split Key™ Cup A.D. with Built-in Validity Drug Screening
Catalog No. 22911070
Change view
Click to view available options
For Use With (Application):
Detects 10 drugs in urine
Detects 5 drugs in urine
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
22-911-070 Detects 5 drugs in urine
22-911-071 Detects 10 drugs in urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-911-070 Supplier Abbott™ Supplier No. DUD157023019
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cup with key activated plunger allows to run device as per convenience
  • Integrated cup screens for testing prescription and illicit drugs in urine
  • Integrated specimen validity combinations available: CR, GL, NI, OX, PH, SG
  • Reserved split-specimen ideal for additional confirmation testing
  • Rapid test results in 5 minutes
Specifications
Detectable Analytes Amphetamine (AMP), Cocaine (COC), Opiates (OPI), Phencyclidine (PCP), Cannabinoids (THC)
Type Integrated E-Z Split Key™ Cup A.D. with Built-in Validity Drug Screening
For Use With (Application) Detects 5 drugs in urine
Quantity 25/Pk.