Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ BRANDplates™ pureGrade™ Strip Plates

Catalog No. p-4954102 Shop All BrandTech Scientific Products
Change view
Click to view available options
Total Volume (Metric) Well:
350 μL
360 μL
Breakapart:
No
Yes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Total Volume (Metric) Well Breakapart
14-380-829 360 μL No
14-380-830 350 μL Yes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-380-829 Supplier BRANDTECH™ BRAND™ Supplier No. 782300
Only null left
Add to Cart
Add to Cart

Non-treated surface strip plates designed to maximize sample recovery and minimize cross-contamination.

  • Medium binding and non-sterile
  • Transparent with flat bottoms
  • Available with or without grid
  • Free from endotoxins, DNase, DNA, RNase, and non-cytotoxic

With grid: Individual strips can be removed from the plate and reinserted. Strips can also be divided into 8 individual vessels and inserted separately into the frame plate.

Without grid: Individual strips can be removed from the frame and reinserted. Strips are not divisible.

Applications: Ideal for homogeneous assays, screening and storage.

TRUSTED_SUSTAINABILITY

Specifications

Barcode Alphanumeric
Breakapart No
Binding Property Untreated
Certified Life Science Quality
Format Strip Plate
Array 12 x 8
Autoclavable Not Autoclavable
Binding Type Medium Binding
Color Clear
Disposable Single-use
Product Type Strip plates
For Use With (Equipment) Microplate readers
Material Polystyrene
Volume (Metric) Well 360 μL
No. of Wells 96
Volume (Metric) Working 360 μL
Sterility Non-sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 360 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) Homogeneous assays, screenings, and storage
Includes 96-well strip plate
Quantity 100/Pk.
RNase Free Yes
Target Molecule Immunoglobulin G (IgG)
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.