Learn More
Hamilton™ 1000 Series Gastight™ Saltline Syringes
Catalog No. 14785628 Shop All Hamilton Company Products
Change view
Click to view available options
Volume (Metric):
1 mL
5 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-785-628 5 mL
14-785-629 10 mL
14-785-627 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-785-628 Supplier Hamilton™ Supplier No. 208336
Add to Cart
Add to Cart
For use with solutions that have a high salt concentration
  • Equipped with inert and non-corrosive plunger
  • Fitted with resistant ultra-high molecular weight poly-ethylene (UHMWPE) plunger tip
Applications:
For life science applications with salty buffers, mining applications with acids or trace metal analysis
Specifications
Autoclavable Not Autoclavable
Product Type Gastight Saltline Syringe
Volume (Metric) 5 mL
Connection Type Bubble Free Prime (BFP)