Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ BRANDplates™ Multiwell Plates for Cell Culture Inserts, - 24-well Standard Plate

Catalog No. 14380865 Shop All BrandTech Scientific Products
Change view
Click to view available options
Surface Treatment:
cellGrade plus
pureGrade S
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment
14-380-865 pureGrade S
14-380-867 cellGrade plus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-380-865 Supplier BRANDTECH™ BRAND™ Supplier No. 782880
Add to Cart
Edge
Add to Cart

Sterile and individually wrapped and available with either untreated or cell culture treated surface.

  • BIO-CERT™ CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, pyrogens, and non-cytotoxic
  • Available with pureGrade™ S and cellGrade™ plus surfaces
  • Includes 24 individually fillable wells that can be fitted with strips of 4 inserts and/or individual inserts
  • Guide ridges for perfect insert fit
  • Designated feeding ports for easy media changes, suitable for robotics/automation
  • 10 plates per pack (individually wrapped with lid)

Surface Information:

  • pureGrade™ S: Non-treated, hydrophobic surface with medium binding capacity (ELISA). Especially suited for bacteriological assays.
  • cellGrade™ plus: Specifically designed for the cultivation of fastidious cell lines. Features carboxyl, hydroxy, and free amino groups on the surface, offering a protein-like composition that facilitates direct cell attachment and spreading.

Applications:

  • Ideal for co-culture, secretions studies, and migrations studies

Compliance

ANSI/SLAS standards 1 and 4

TRUSTED_SUSTAINABILITY

Specifications

Closure Included Yes
Bottom Shape Flat
Capacity (Metric) 3.1 mL
Color Transparent
Culture Area 210 mm2
Material Polystyrene
Sterility Sterile
Surface Treatment pureGrade S
Type Cell Culture Insert System
Well Count 24
For Use With (Application) Co-culture, secretion studies, migration studies
For Use With (Equipment) Insert system
Height (Metric) 19.5 mm
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.