Learn More
BRANDTECH™ BRAND™ BRANDplates™ Multiwell Plates for Cell Culture Inserts, - 6-well Specialized Plate
Shop All BRANDTECH Scientific ProductsDescription
- BIO-CERT™ CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, pyrogens, and non-cytotoxic
- Available with pureGrade™ S and cellGrade™ plus surfaces
- 6-well special plate design connects 4 wells into one large, elongated well, allowing simultaneous medium supply to all 4 inserts in a strip
- Ideal for insert strips with inlet channels, it also supports single inserts and combinations of 2 or 3 connected inserts
- Guide ridges for perfect insert fit
- Designated feeding ports for easy media changes, suitable for robotics/automation
- 10 plates per pack (individually wrapped with lid)
Surface Information:
- pureGrade™ S: Non-treated, hydrophobic surface with medium binding capacity (ELISA). Especially suited for bacteriological assays.
- cellGrade™ plus: Specifically designed for the cultivation of fastidious cell lines. Features carboxyl, hydroxy, and free amino groups on the surface, offering a protein-like composition that facilitates direct cell attachment and spreading.
Applications:
- Ideal for 3D tissue cultures
Compliance
ANSI/SLAS standards 1 and 4
Specifications
Specifications
| Closure Included | Yes |
| Bottom Shape | Flat |
| Capacity (Metric) | 10 mL |
| Color | Transparent |
| Culture Area | 855 mm2 |
| Material | Polystyrene |
| Sterility | Sterile |
| Surface Treatment | cellGrade plus |
| Type | Cell Culture Insert System |
| Well Count | 6 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.