Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BASIC™ Universal Pads

Catalog No. 17111158 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
17 gal./Bale
20.5 gal./Bale
34 gal./Bale
Weight Capacity:
Heavyweight
Lightweight
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Weight Capacity
17-111-158 34 gal./Bale Lightweight
17-111-145 17 gal./Bale Lightweight
17-111-144 20.5 gal./Bale Heavyweight
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-111-158 Supplier Brady™ Supplier No. BPU200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

General purpose sorbent to clean up oils, coolants, solvents, and water-based fluids

  • Sorbents absorb up to 20 times their weight
  • Single-ply meltblown polypropylene
  • Perforated and dimpled
  • Absorbencies: Heavy and light weight
  • Size: 38.1 x 43.1cm (15 x 17 in.)
  • Specifications

    Color Gray
    Capacity (English) 34 gal./Bale
    Material Polypropylene
    Type Universal Absorbent Pad
    For Use With (Application) Clean up oils, coolants, solvents, and water-based fluids
    Model BASIC Series
    Packaging Quantity 200/Cs.
    Weight Capacity Lightweight
    Dimensions (L x W) 38.1 x 43.1 cm (15 x 17 in.)
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.