Learn More
Brady™ ENV™ Marine Sorbent Booms
Catalog No. 17111089 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
32 gal./Bale
65 gal./Bale
Dimensions (L x W):
10 ft. x 5 in.
10 ft. x 8 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Dimensions (L x W)
17-111-089 32 gal./Bale 10 ft. x 5 in.
17-111-091 65 gal./Bale 10 ft. x 8 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-111-089 Supplier Brady™ Supplier No. ENV510
  • $272.00
    $199.65 / Case of 4
    Save $72.35
    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Designed to float on water and contain or absorb wide variety of marine fuels and oils
  • Feature lint-free blue sleeve
  • Class: Oil only
  • Available in two sizes
  • Specifications
    Capacity (English) 32 gal./Bale
    Material Polypropylene
    Type Sorbent Boom
    Packaging Quantity 4/Cs.
    Dimensions (L x W) 10 ft. x 5 in.