Learn More
Brady™ ENV™ Marine Sorbent Booms
Catalog No. 17111091 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
32 gal./Bale
65 gal./Bale
Dimensions (L x W):
10 ft. x 5 in.
10 ft. x 8 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Dimensions (L x W)
17-111-091 65 gal./Bale 10 ft. x 8 in.
17-111-089 32 gal./Bale 10 ft. x 5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-111-091 Supplier Brady™ Supplier No. ENV810 /DEL
Designed to float on water and contain or absorb wide variety of marine fuels and oils
  • Feature lint-free blue sleeve
  • Class: Oil only
  • Available in two sizes
  • Specifications
    Capacity (English) 65 gal./Bale
    Type Sorbent Boom
    Packaging Quantity 4/Cs.
    Dimensions (L x W) 10 ft. x 8 in.