Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ High Traffic Series Medium Weight Rolls

Catalog No. 17111107 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
32 gal.
32 gal./Bale
65 gal.
Dimensions (L x W):
150 ft. x 30 in.
300 ft. x 15 in.
300 ft. x 30 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Dimensions (L x W)
17-111-107 32 gal./Bale 150 ft. x 30 in.
17-111-110 65 gal. 300 ft. x 30 in.
17-111-104 32 gal. 300 ft. x 15 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-111-107 Supplier Brady™ Supplier No. HT350
Add to Cart
Add to Cart

Made to withstand heavy foot and forklift traffic along with problem spill areas

  • Features gray coverstock to mask spills
  • Universal 2-ply construction
  • Upper layer is abrasion-resistant
  • Perforated
  • Available in various sizes
  • Specifications

    Color Gray
    Capacity (English) 32 gal./Bale
    Material Polypropylene
    Type High Traffic Series Medium-weight Roll
    For Use With (Application) Walkways, overspray leaks and drips, workstations, work surfaces and under assembly lines
    Packaging Quantity 1 Ea.
    Weight Capacity Medium-weight
    Dimensions (L x W) 150 ft. x 30 in.
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.