Learn More
Brady™ MRO Plus™ Universal Heavy Pads
Catalog No. 17111162 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
13.5 gal./Bale
26 gal.
41 gal./Bale
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17-111-162 13.5 gal./Bale
17-111-151 26 gal.
17-111-163 41 gal./Bale
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-111-162 Supplier Brady™ Supplier No. MRO1002
Add to Cart
Add to Cart
Absorbs all industrial liquids (water, petroleum-based and chemical-based fluids)
  • Three-ply construction
  • Dimpled and Perforated - reduce waste
  • Gray color masks drips and spills
  • Specifications
    Color Gray
    Capacity (English) 13.5 gal./Bale
    Material Polypropylene
    Type Universal Absorbent Pad
    For Use With (Application) Overspray leaks and drips, workstations, heavy clean-up and wipe-up, drums - liquid storage, outfalls and discharge ponds
    Packaging Quantity 50/Cs.
    Dimensions (L x W) 38 x 48.26 cm (15 x 19 in.)
    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov