Learn More
Brady™ Oil Plus™ Heavy Pad
Catalog No. 17111129 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
26 gal./Bale
41 gal./Bale
Dimensions (L x W):
38.1 x 48.2 cm (15 x 19 in.)
76.2 x 76.2 cm (30 x 30 in.)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Dimensions (L x W)
17-111-129 26 gal./Bale 38.1 x 48.2 cm (15 x 19 in.)
17-111-143 41 gal./Bale 76.2 x 76.2 cm (30 x 30 in.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-111-129 Supplier Brady™ Supplier No. OP100
Add to Cart
Add to Cart
Absorbs petroleum-based oils, solvents and other non-water soluble liquids
  • Floats indefinitely even when saturated with oil
  • 3-ply (MMM) Meltblown Polypropylene construction
  • Dimpled and perforated
  • Available in various sizes
  • Specifications
    Color White
    Capacity (English) 26 gal./Bale
    Material Polypropylene
    Type Petroleum Sorbent Pad
    For Use With (Application) Overspray leaks and drips, workstations, heavy clean-up and wipe-up , drums - liquid storage, outfalls and discharge ponds
    Packaging Quantity 100/Cs.
    Weight Capacity Heavyweight
    Dimensions (L x W) 38.1 x 48.2 cm (15 x 19 in.)