Learn More
Brady™ Oil Plus™ Heavy Pad
Catalog No. 17111143 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
26 gal./Bale
41 gal./Bale
Dimensions (L x W):
38.1 x 48.2 cm (15 x 19 in.)
76.2 x 76.2 cm (30 x 30 in.)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Dimensions (L x W)
17-111-143 41 gal./Bale 76.2 x 76.2 cm (30 x 30 in.)
17-111-129 26 gal./Bale 38.1 x 48.2 cm (15 x 19 in.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-111-143 Supplier Brady™ Supplier No. OP50
  • $248.00
    $180.65 / Case of 50
    Save $67.35
    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Absorbs petroleum-based oils, solvents and other non-water soluble liquids
  • Floats indefinitely even when saturated with oil
  • 3-ply (MMM) Meltblown Polypropylene construction
  • Dimpled and perforated
  • Available in various sizes
  • Specifications
    Color White
    Capacity (English) 41 gal./Bale
    Material Polypropylene
    Type Petroleum Sorbent Pad
    For Use With (Application) Overspray leaks and drips, workstations, heavy clean-up and wipe-up , drums - liquid storage, outfalls and discharge ponds
    Packaging Quantity 50/Cs.
    Weight Capacity Heavyweight
    Dimensions (L x W) 76.2 x 76.2 cm (30 x 30 in.)