Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ SPC™ Rag Rug Absorbent Mats

Catalog No. 17111117 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
37 gal.
38 gal.
Dimensions (L x W):
150 ft. x 24 in.
45.7 x 45.7 cm (18 x 18 in.)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Dimensions (L x W)
17-111-117 38 gal. 45.7 x 45.7 cm (18 x 18 in.)
17-111-100 37 gal. 150 ft. x 24 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-111-117 Supplier Brady™ Supplier No. RAG1818
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provides good footing, masks dirt and absorbs spills

  • Made of cotton and polyester fibers
  • Soak up oil, water-based and solvent-based liquids
  • For maintenance, repair and industrial environments
  • Available in various sizes

Specifications

Color Gray
Capacity (English) 38 gal.
Material Cotton, Polyester
Type Rag Rug Absorbent Mat
For Use With (Application) Overspray leaks and drips, workstations, drums - liquid storage, outfalls and discharge ponds
Dimensions (L x W) 45.7 x 45.7 cm (18 x 18 in.)
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.