Learn More
Brady™ SXT Oil Only Absorbent Pads
Catalog No. 17111132 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
17 gal./Bale
19.5 gal./Bale
31.5 gal./Bale
Weight Capacity:
Heavyweight
Lightweight
Medium-weight
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Weight Capacity
17-111-132 17 gal./Bale Medium-weight
17-111-137 31.5 gal./Bale Lightweight
17-111-131 19.5 gal./Bale Heavyweight
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-111-132 Supplier Brady™ Supplier No. SXT300
  • $111.00
    $78.65 / Case of 100
    Save $32.35
    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Made to handle petroleum-based spills
  • Features three Ply Spunbond Coverstock on both sides for low linting
  • Abrasion-resistance and non-abrasive performance
  • Remains intact even when saturated
  • For workstations involving the wipe down of tools or component parts, wringing and re-using of sorbents
  • Perforated - reduces waste
  • Specifications
    Capacity (English) 17 gal./Bale
    Material Polypropylene
    Type Absorbent Pad, Oil-only
    For Use With (Application) Overspray leaks and drips, workstations, heavy clean-up and wipe-up , drums - liquid storage, outfalls and discharge ponds, food processing, high tech industries, machining of small parts, paper industry, pharmaceutical
    Packaging Quantity 100/Cs.
    Weight Capacity Medium-weight
    Dimensions (L x W) 38.1 x 48.2 cm (15 x 19 in.)
    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov