Learn More
Brady™ SXT Oil Only Heavy Rolls
Catalog No. 17111188 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
18 gal./Bale
37 gal.
Dimensions (L x W):
150 ft. x 15 in.
150 ft. x 30 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Dimensions (L x W)
17-111-188 37 gal. 150 ft. x 30 in.
17-111-181 18 gal./Bale 150 ft. x 15 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-111-188 Supplier Brady™ Supplier No. SXT30DP
  • $240.00
    $169.65 / Each of 1
    Save $70.35
    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Oil only sorbent made to handle petroleum-based spills
  • Remains intact even when saturated
  • Spunbond coverstock on both sides
  • Ideal for workstations involving the wipe down of tools or component parts
  • Perforated
  • Specifications
    Capacity (English) 37 gal.
    Material Polypropylene
    Type Oil Only Heavy Roll
    For Use With (Application) Food Processing, High Tech Industries, Machining of small parts, Paper Industry, Pharmaceutical
    Packaging Quantity 1 Ea.
    Weight Capacity Heavyweight
    Dimensions (L x W) 150 ft. x 30 in.