Learn More
Brady™ TrackMat™ Heavy Roll Absorbents
Catalog No. 17111097 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
20 gal./Bale
26 gal./Bale
39 gal./Bale
Dimensions (L x W):
80 ft. x 19 in.
80 ft. x 29 in.
80 ft. x 58 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Dimensions (L x W)
17-111-097 26 gal./Bale 80 ft. x 19 in.
17-111-096 39 gal./Bale 80 ft. x 58 in.
17-111-095 20 gal./Bale 80 ft. x 29 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-111-097 Supplier Brady™ Supplier No. TM19
  • $470.00
    $332.65 / Case of 2
    Save $137.35
    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Barrier sorbent for high volume petroleum spills
  • Three-ply construction
  • Features UV-resistant top layer, absorbent polypropylene core, and non-leaking and non-slip barrier backing
  • Fits outside standard train rails
  • Available in various sizes
Specifications
Capacity (English) 26 gal./Bale
Type TrackMat Heavy Roll Absorbent
For Use With (Application) Overspray leaks and drips, workstations, drums - liquid storage, outfalls and discharge ponds, refineries, military, utilities, steel mills
Packaging Quantity 2/Cs.
Weight Capacity Heavyweight
Dimensions (L x W) 80 ft. x 19 in.