Learn More
Brady™ TrackMat™ Heavy Roll Absorbents
Catalog No. p-4955335 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
20 gal./Bale
26 gal./Bale
39 gal./Bale
Dimensions (L x W):
80 ft. x 19 in.
80 ft. x 29 in.
80 ft. x 58 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Dimensions (L x W)
17-111-097 26 gal./Bale 80 ft. x 19 in.
17-111-096 39 gal./Bale 80 ft. x 58 in.
17-111-095 20 gal./Bale 80 ft. x 29 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-111-097 Supplier Brady™ Supplier No. TM19
Only null left
Add to Cart
Add to Cart
Barrier sorbent for high volume petroleum spills
  • Three-ply construction
  • Features UV-resistant top layer, absorbent polypropylene core, and non-leaking and non-slip barrier backing
  • Fits outside standard train rails
  • Available in various sizes
Specifications
Capacity (English) 26 gal./Bale
Type TrackMat Heavy Roll Absorbent
For Use With (Application) Overspray leaks and drips, workstations, drums - liquid storage, outfalls and discharge ponds, refineries, military, utilities, steel mills
Packaging Quantity 2/Cs.
Weight Capacity Heavyweight
Dimensions (L x W) 80 ft. x 19 in.