Learn More
Brady™ Cansorb™ Oil Only Granular Sorbents
Catalog No. 17111077 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
10 gal.
Type:
Loose Granular Sorbent
Catalog No. Capacity (English) Type
17-111-077 10 gal. Loose Granular Sorbent
1 options
Catalog No. 17-111-077 Supplier Brady™ Supplier No. CSLFB10
Ideal for oil-only clean up on open water and land applications
  • Made from fiber sphagnum peat moss
  • Class: Oil only
  • Absorbs 8 times its weight
  • Available in compressed and loose granular format
  • Specifications
    Capacity (English) 10 gal.
    Capacity (Metric) 37.85 L
    Material Fiber Sphagnum Peat Moss
    Type Loose Granular Sorbent
    Size 10