Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

InVitroCare™ Sperm Wash Media

Catalog No. 23000904
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
12 x 12 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-000-904 100 mL
23-000-903 12 x 12 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-000-904 Supplier Invitro Care Inc Supplier No. 2005
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For in vitro sperm washing procedures

  • HEPES buffered formulation with 0.5% Human Serum Albumin
  • Does not require use of CO2 incubator
  • Shelf life: 18 months

Specifications

Content And Storage 2°-8°C
Product Type Sperm Wash Medium
With Additives HEPES
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.