Learn More
Justrite™ Floor Grating w/Sump Liner
Catalog No. 19414033
Change view
Click to view available options
For Use With (Equipment):
12-drum/387 cu. ft. Lockers
16-drum/506 cu. ft. Lockers
2-drum/ (52 cu. ft.) lockers
4-drum/ (105 cu. ft.) lockers
6-drum/ (205 cu. ft.) lockers
9-drum/ (296 cu. ft.) lockers
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-414-033 16-drum/506 cu. ft. Lockers
19-414-032 12-drum/387 cu. ft. Lockers
19-414-030 6-drum/ (205 cu. ft.) lockers
19-414-031 9-drum/ (296 cu. ft.) lockers
19-414-029 4-drum/ (105 cu. ft.) lockers
19-414-028 2-drum/ (52 cu. ft.) lockers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-414-033 Supplier Justrite™ Supplier No. 915211
Add to Cart
Add to Cart
Request a Quote
Accessory for Outdoor Safety Lockers
  • For use with 2-,4-,6-,9-,12-,16-drum lockers
  • Made of High density polyethylene (HDPE) sump liner
  • Fiberglass grating
Applications:
For acid and corrosive chemical spill resistance applications
Specifications
Product Type Floor Grating
For Use With (Equipment) 16-drum/506 cu. ft. Lockers
Material HDPE
Description HDPE sump liner
Includes Sump liner