Learn More
Justrite™ Explosion-Proof Heaters
Catalog No. 19414035 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
12-,16-drum fire rated Agri-Turf lockers
6-,9-drum fire rated Agri-Turf lockers
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-414-035 12-,16-drum fire rated Agri-Turf lockers
19-414-034 6-,9-drum fire rated Agri-Turf lockers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-414-035 Supplier Justrite™ Supplier No. 915303
Accessory for Outdoor Safety Lockers
  • Wall mounted
  • For use with 6-,9-,12-,16-drum fire-rated Agri-Turf™ lockers only
  • Available in 6000 BTU and 12000 BTU heating capacity
Applications:
Adds heating where comfort, chemical stability or freeze protection is important
Certifications
UL listed
†For optimum temperature control, appropriate size R-11 insulation package must be ordered when unit is purchased in conjunction with fire-rated models. This accessory must be purchased in conjunction with locker to ensure proper installation and to retain FM-approval.
Specifications
Product Type Explosion-proof heater
For Use With (Equipment) 12-,16-drum fire rated Agri-Turf lockers