Learn More
Cellucap Spunbond Polypropylene Coveralls, Elastic Wrists and Ankles
Catalog No. 19133906 Shop All Cellucap Products
Change view
Click to view available options
Size:
Small
Medium
Large
3X-Large
4X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-133-906 4X-Large
19-133-905 3X-Large
19-133-902 Large
19-133-900 Small
19-133-901 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-133-906 Supplier Cellucap Supplier No. 55840XXXX
Add to Cart
Add to Cart
This item is not returnable. View return policy
Coveralls constructed of non-woven material offer lightweight splash protection from non-hazardous liquids and dry particulates.
  • Non-woven spunbond polypropylene
  • Lightweight splash protection from non-hazardous liquids and dry particulates
  • 1.25 oz. material
  • Elastic wrists and ankles
  • Collared
Specifications
Product Type General Purpose Coveralls
Size 4X-Large
Gender Unisex
Material Spunbond Polypropylene
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Liquid, Particle
Description 1.25 oz. Spunbound Polypropylene
Model 55840