Learn More
Justrite™ Explosion-Proof Air Conditioners
Catalog No. 19414037 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
12-,16-drum fire rated Agri-Turf lockers
6-,9-drum fire rated Agri-Turf lockers
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-414-037 12-,16-drum fire rated Agri-Turf lockers
19-414-036 6-,9-drum fire rated Agri-Turf lockers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-414-037 Supplier Justrite™ Supplier No. 915307
  • $46,357.00
    $31,580.65 / Each of 1
    Save $14,776.35
    32% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Request a Quote
Accessory for Outdoor Safety Lockers
  • For use with 6-,9-,12-,16-drum fire-rated Agri-Turf™ lockers only
  • Available in 4.4kWh (15000 BTU) and 5.9kWh (20000 BTU) energy capacity
Applications:
Adds air conditioning where comfort, chemical stability or freeze protection is important
†For optimum temperature control, appropriate size R-11 insulation package must be ordered when unit is purchased in conjunction with fire-rated models. This accessory must be purchased in conjunction with locker to ensure proper installation and to retain FM-approval.
Specifications
Product Type Explosion-proof Air conditioner
For Use With (Equipment) 12-,16-drum fire rated Agri-Turf lockers