Learn More
Justrite™ Explosion-Proof Heater/Air-Conditioner Package
Catalog No. 19414038 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
12-,16-drum fire rated Agri-Turf lockers
6-,9-drum fire rated Agri-Turf lockers
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-414-038 6-,9-drum fire rated Agri-Turf lockers
19-414-039 12-,16-drum fire rated Agri-Turf lockers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-414-038 Supplier Justrite™ Supplier No. 915309
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Accessory for Outdoor Safety Lockers
  • For use with 6-,9-,12-,16-drum fire-rated Agri-Turf™ lockers only
  • Available in 1.7kWh (6000 BTU) and 3.51kWh (12000 BTU) heater
  • 4.4kWh (15000 BTU) and 5.9kWh (20000 BTU) air conditioner
Applications:
Adds heating/air conditioning where comfort, chemical stability or freeze protection is important
†For optimum temperature control, appropriate size R-11 insulation package must be ordered when unit is purchased in conjunction with fire-rated models. This accessory must be purchased in conjunction with locker to ensure proper installation and to retain FM-approval.
Specifications
Product Type Heater/Air Conditioner Package
For Use With (Equipment) 6-,9-drum fire rated Agri-Turf lockers