Learn More
Tians epic™ Polypropylene White Coveralls
Catalog No. 19168955 Shop All Tians International Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-168-955 -
19-168-953 X-Large
19-168-952 Large
19-168-951 -
19-168-950 -
19-168-954 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-168-955 Supplier Tians Supplier No. 2108813XL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Coverall with elastic wrists and zipper front
  • Made of breathable polypropylene fabric
  • ISO Class 7 compatible
  • Available in different sizes
  • White in color
Specifications
Material Polypropylene
Color White