Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sundance Solar™ Sunbender Do-It-Yourself Solar LED Jar Light Kit-Pre Wired
SCI_ED

Catalog No. p-4961271
Encompass
Change view
Click to view available options
Color:
Blue
White
Catalog No. Color
S12854 Blue
S12853 White
2 options
This item is not returnable. View return policy

Catalog No. S12854

Supplier: Sundance Solar™ 9001001735BL

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This item is not returnable. View return policy

Helps build colorful LED light that automatically comes on at night and goes off at dawn.

  • Made of 3V 70mA polycrystalline solar panel
  • Has dusk/dawn circuit board
  • Has two bright 5mm LEDs
  • Has 2 AAA NiMH rechargeable batteries
  • Battery holder and wires, wire nuts

Specifications

Amperage 70 mA
Battery Type AAA NiMH rechargeable batterie
Color Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.