Learn More
Medchemexpress LLC 4a-Phorbol 12 13-did 1mg | 27536-56-7 | 672.93 g·mol⁻1 | C40H64O8 | 1 MG
SDP
Catalog No. 5000308055
Encompass_Preferred
4α-Phorbol 12,13-didecanoate is a phorbol ester research reagent used as a selective activator of the TRPV4 ion channel. It is employed to study TRPV4-mediated calcium influx, osmoregulatory responses, and related signaling pathways, and is supplied in small, high-purity laboratory quantities.
  • Selective TRPV4 agonist for calcium influx studies
  • High purity, typically 99.3%
  • Supplied in small laboratory quantities suitable for in vitro research (for example, 1 mg)
  • Applicable to studies of osmoregulation and membrane transporter signaling
Catalog No. 50-003-08055 Supplier Medchemexpress LLC Supplier No. HY1162911MG
Add to Cart
Add to Cart
4α-Phorbol 12,13-didecanoate is a phorbol ester research reagent used as a selective activator of the TRPV4 ion channel. It is employed to study TRPV4-mediated calcium influx, osmoregulatory responses, and related signaling pathways, and is supplied in small, high-purity laboratory quantities.
  • Selective TRPV4 agonist for calcium influx studies
  • High purity, typically 99.3%
  • Supplied in small laboratory quantities suitable for in vitro research (for example, 1 mg)
  • Applicable to studies of osmoregulation and membrane transporter signaling