Learn More
Research Products International Corp 4a20 Complete Counting Cocktail, 4 Liters
SDP
Supplier:  Research Products International Corp 111160
Encompass_Preferred
Complete Counting Cocktail 4a20 is a ready to use cocktail for liquid scintillation counting. It contains a blend of PPO and bis-MSB in Xylene (Scintillation Grade) yielding excellent quench resistance and high counting efficiency and will handle a wide range of non-aqueous samples. May also be used as an extractant and counting solution for organic samples separated by TLC or on filter discs.
Catalog No. 50-197-7630