Learn More
Cayman Chemical 4A7C-301-Nurr1 Agonist, Purity: ≥98%, 5 mg
SDP
Catalog No. 502917001
Encompass_Preferred
A Nurr1 receptor agonist; binds to Nurr1-LBD (IC50 = 48.22 nM) and increases transcriptional activity of Nurr1-LBD and full-length Nurr1 in reporter assays using SK-N-BE(2)C human neuroblastoma cells (EC50s = 6.53 and 50-70 µM, respectively); decreases dopaminergic cell death in the striatum and substantia nigra pars compacta, as well as reduces motor and olfactory deficits, without inducing dyskinesia-like behaviors in MPTP- or α-synuclein-overexpression mouse models of Parkinson’s disease
Catalog No. 50-291-7001 Supplier Cayman Chemical Supplier No. 394535 MG
A Nurr1 receptor agonist; binds to Nurr1-LBD (IC50 = 48.22 nM) and increases transcriptional activity of Nurr1-LBD and full-length Nurr1 in reporter assays using SK-N-BE(2)C human neuroblastoma cells (EC50s = 6.53 and 50-70 µM, respectively); decreases dopaminergic cell death in the striatum and substantia nigra pars compacta, as well as reduces motor and olfactory deficits, without inducing dyskinesia-like behaviors in MPTP- or α-synuclein-overexpression mouse models of Parkinson’s disease