Learn More
Medchemexpress LLC HY-129434A 5mg , 4aα,7α,7aα-Nepetalactone CAS:21651-62-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY129434A5MG
Encompass_Preferred
HY-129434A 5mg 4aα,7α,7aα-Nepetalactone CAS: 21651-62-7 4aα,7α,7aα-Nepetalactone exhibits antibacterial activity, and inhibits Escherichia coli, Pseudomonas aeruginosa, Staphylococcus aureus, Salmonella typhi and Enterococcus faecalis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1744