Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ 4AA ProPolymer LED Flashlight

Catalog No. 19819860 Shop All Streamlight Products
Change view
Click to view available options
Light Source:
High Power LED
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Light Source
19-819-860 High Power LED
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-819-860 Supplier Streamlight™ Supplier No. 68200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The 4AA ProPolymer is an intrinsically safe polymer flashlight with extended runtime for any industrial lighting needs that require a soft flood light. Class I, Div 1 rated for use in hazardous environments

  • Class 1 Div. 1 rated
  • Runtime 155 hours
  • Runs on four AA alkaline batteries, not included
  • Comfortable 6.5 in. size and ergonomically shaped for hands
  • Push button tail switch for one handed operation
  • Waterproof and made of impact resistant polymer with a rubber face cap
  • Non-conductive and corrosion proof

Specifications

Light Source High Power LED
Lighting Life 100,000 hr.
Weight (English) 6.3 oz.
Weight (Metric) 178.6 g
Battery Run Time 155 hours
Battery Type AA Alkaline
Certifications/Compliance Class I, Division 1; Class I, II, Division 2; Class III
Color Yellow
Lens Material Polycarbonate
Lumen 67 lm
Warranty Limited lifetime warranty
Product Type Handheld Flashlight
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.