Learn More
Cayman Chemical 4-acetoxy DMT (hydrochloride), Purity: ≥90%, CAS: 2748484-99-1, 10 mg
SDP
Catalog No. 502953801
Encompass_Preferred
An analytical reference standard categorized as a tryptamine; a prodrug form of psilocin; induces the HTR in mice, indicating hallucinogenic potential; intended for research and forensic applications
Catalog No. 50-295-3801 Supplier Cayman Chemical Supplier No. 1405610 MG
An analytical reference standard categorized as a tryptamine; a prodrug form of psilocin; induces the HTR in mice, indicating hallucinogenic potential; intended for research and forensic applications