Learn More
Cayman Chemical 4acetoxy METhydrochLRIde 10mg
SDP
Catalog No. 502967056
Encompass_Preferred
An analytical reference standard categorized as a tryptamine; induces the HTR in mice; intended for research and forensic applications
Catalog No. 50-296-7056 Supplier Cayman Chemical Supplier No. 1836710 MG
An analytical reference standard categorized as a tryptamine; induces the HTR in mice; intended for research and forensic applications