Learn More
Medchemexpress LLC 4alpha-phorbol 12-myristate 13-acetate | 63597-44-4 | MFCD00153860 | 99.0% | 616.83 | C36H56O8 | 1 MG
SDP
Catalog No. 5000242922
Encompass_Preferred
4alpha-phorbol 12-myristate 13-acetate is the inactive stereoisomer of TPA used as a negative control in experiments that monitor TPA-activated signaling events. It is supplied as a high-purity research reagent in a small-mass vial suitable for in vitro assays.
  • Inactive negative control for TPA-activated events.
  • High purity (99.0%) for reproducible results.
  • Molecular formula C36H56O8 and molecular weight 616.83 for accurate identification.
  • Supplied in a 1 mg package suitable for small-scale studies.
Catalog No. 50-002-42922 Supplier Medchemexpress LLC Supplier No. HY1060291MG
4alpha-phorbol 12-myristate 13-acetate is the inactive stereoisomer of TPA used as a negative control in experiments that monitor TPA-activated signaling events. It is supplied as a high-purity research reagent in a small-mass vial suitable for in vitro assays.
  • Inactive negative control for TPA-activated events.
  • High purity (99.0%) for reproducible results.
  • Molecular formula C36H56O8 and molecular weight 616.83 for accurate identification.
  • Supplied in a 1 mg package suitable for small-scale studies.