Learn More
Cayman Chemical 4-Amino-5-(bromomethyl)-2-methylpyrimidine (hydrobromide), Purity: ≥95%, CAS: 5423-98-3, 10 mg
SDP
Catalog No. 502988290
Encompass_Preferred
A heterocyclic building block; has been used in the synthesis of vitamin B1 analogs and thiaminephosphoric acid esters
Catalog No. 50-298-8290 Supplier Cayman Chemical Supplier No. 2983910 MG
A heterocyclic building block; has been used in the synthesis of vitamin B1 analogs and thiaminephosphoric acid esters