Learn More
Cayman Chemical 4-Aminobenzylphosphonic Acid, Purity: ≥95%, CAS: 5424-27-1, 250 mg
SDP
Catalog No. 502912649
Encompass_Preferred
An alkaline phosphatase inhibitor (Ki = 1.1 mM for the bovine intestine enzyme); has been used in affinity chromatography for the purification of alkaline phosphatase; has been used in electrochemical modification of glassy carbon electrodes
Catalog No. 50-291-2649 Supplier Cayman Chemical Supplier No. 36029250 MG
An alkaline phosphatase inhibitor (Ki = 1.1 mM for the bovine intestine enzyme); has been used in affinity chromatography for the purification of alkaline phosphatase; has been used in electrochemical modification of glassy carbon electrodes