Learn More
Cayman Chemical 4AmInomethyl4 5 8trImeth 10mg
SDP
Catalog No. 502965022
Encompass_Preferred
A bifunctional photoreactive agent for crosslinking nucleic acids; intercalates into double-stranded regions in DNA or RNA and, upon UV irradiation, forms covalent adducts with pyrimidine bases; used to study different types of RNA from diverse organisms, including viruses
Catalog No. 50-296-5022 Supplier Cayman Chemical Supplier No. 1716210 MG
A bifunctional photoreactive agent for crosslinking nucleic acids; intercalates into double-stranded regions in DNA or RNA and, upon UV irradiation, forms covalent adducts with pyrimidine bases; used to study different types of RNA from diverse organisms, including viruses