Learn More
Cayman Chemical 4AmInophenyl PhosphatSodI 10mg
SDP
Catalog No. 502965131
Encompass_Preferred
A substrate for alkaline phosphatase that can be used in enzymatic reactions to quantify ALP activity
Catalog No. 50-296-5131 Supplier Cayman Chemical Supplier No. 1729310 MG
A substrate for alkaline phosphatase that can be used in enzymatic reactions to quantify ALP activity