Learn More
Cayman Chemical 4-Aminophenyl Phosphate (sodium salt hydrate), Purity: ≥95%, 50 mg
SDP
Catalog No. 502965716
Encompass_Preferred
A substrate for alkaline phosphatase that can be used in enzymatic reactions to quantify ALP activity
Catalog No. 50-296-5716 Supplier Cayman Chemical Supplier No. 1729350 MG
A substrate for alkaline phosphatase that can be used in enzymatic reactions to quantify ALP activity