Learn More
Cayman Chemical 4-Aminophenylphosphorylcholine, Purity: ≥95%, CAS: 102185-28-4, 5 mg
SDP
Catalog No. 502982967
Encompass_Preferred
A probe that can be conjugated to sepharose in order to generate an adsorbent for use in affinity precipitation experiments
Catalog No. 50-298-2967 Supplier Cayman Chemical Supplier No. 206435 MG
A probe that can be conjugated to sepharose in order to generate an adsorbent for use in affinity precipitation experiments