Learn More
Cayman Chemical 4AMNoPHNylphsphorylcholn 10mg
SDP
Catalog No. 502984731
Encompass_Preferred
A probe that can be conjugated to sepharose in order to generate an adsorbent for use in affinity precipitation experiments
Catalog No. 50-298-4731 Supplier Cayman Chemical Supplier No. 2064310 MG
A probe that can be conjugated to sepharose in order to generate an adsorbent for use in affinity precipitation experiments